CLS3620
Corning® Costar® microcentrifuge tubes with snap cap
1.7 mL snap cap microcentrifuge tube, polypropylene, non-sterile, natural, 500/cs
Synonym(s):
centrifuge tubes, microcentrifuge tubes, microfuge tubes, snap cap microcentrifuge tubes
About This Item
Recommended Products
material
conical bottom tube
polypropylene cap
polypropylene tube
sterility
non-sterile
feature
RCF 17,000 × g
RNase and DNase free
autoclavable
disposable
graduations
self-standing: no (style: (conical bottom))
packaging
case of 500
pack of 500
manufacturer/tradename
Corning 3620
parameter
17000 × g max. RCF
cap diam.
0.362 in.
capacity
1.7 mL
tube O.D.
.500 in.
color
natural
fitting
cap (push fit)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Supplied nonsterile and are autoclavable
- External graduations and frosted writing spot for easy sample identification
- Positive seal design allows for repeated opening and closing
- Flat cap surface for convenient labeling
- Withstands a maximum RCF of 17,000 × g
- Assorted color pack includes one bag each of blue, green, yellow, red and orange tubes.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service