Skip to Content
Merck
All Photos(1)

Key Documents

602590

Sigma-Aldrich

5,5,5-Trifluoro-L-leucine, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18F3NO4
Molecular Weight:
285.26
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

form

solid

mol wt

285.26 by atom % calculation

application(s)

peptide synthesis

functional group

Boc

SMILES string

CC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)C(F)(F)F

InChI

1S/C11H18F3NO4/c1-6(11(12,13)14)5-7(8(16)17)15-9(18)19-10(2,3)4/h6-7H,5H2,1-4H3,(H,15,18)(H,16,17)/t6?,7-/m0/s1

InChI key

PLTXZUYAJDKNFI-MLWJPKLSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service