Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3138020
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
[H][C@]12CCC(C)=C1[C@@H](CC[C@H]2C)\C=C(/C)C(O)=O
InChI
1S/C15H22O2/c1-9-4-6-12(8-11(3)15(16)17)14-10(2)5-7-13(9)14/h8-9,12-13H,4-7H2,1-3H3,(H,16,17)/b11-8+/t9-,12+,13-/m1/s1
InChI key
FEBNTWHYQKGEIQ-SUKRRCERSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade primary reference standard | grade analytical standard | grade primary reference standard |
| manufacturer/tradename USP | manufacturer/tradename PhytoLab | manufacturer/tradename - | manufacturer/tradename HWI |
| application(s) pharmaceutical (small molecule) | application(s) - | application(s) cleaning products | application(s) food and beverages |
| format neat | format - | format neat | format - |
Still not finding the right product?
Explore all of our products under Valerenic acid
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1707908-30MG | 04061834091417 |
