Skip to Content
Merck
All Photos(1)

Key Documents

578495

Sigma-Aldrich

Methyl indole-6-carboxylate

97%

Synonym(s):

Indole-6-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

76-80 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc2cc[nH]c2c1

InChI

1S/C10H9NO2/c1-13-10(12)8-3-2-7-4-5-11-9(7)6-8/h2-6,11H,1H3

InChI key

AYYOZKHMSABVRP-UHFFFAOYSA-N

Application

Reactant for preparation of:
Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
Inhibitor of botulinum neurotoxin
ITK inhibitors
Antibacterial agents
CB2 cannabinoid receptor ligands
Inhibitors of hepatitis C virus NS5B polymerase
Inhibitors with histamine H1-blocking activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Heine et al.
Applied biochemistry and biotechnology, 181(4), 1590-1610 (2016-11-11)
The enantioselective epoxidation of styrene and related compounds by two-component styrene monooxygenases (SMOs) has targeted these enzymes for development as biocatalysts. In the present work, we prepare genetically engineered fusion proteins that join the C-terminus of the epoxidase (StyA) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service