Principaux documents
TMO312751-9050
Nalgene® Dropper Bottle without tip or cap
white low-density polyethylene, volume 15 mL, case of 2000 ea
Synonyme(s) :
LDPE dropper bottle, dispensing bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
white low-density polyethylene
Conditionnement
case of 2000 ea
Fabricant/nom de marque
Nalgene 312751-9050
Capacité
15 mL
Volume
15 mL
Description générale
Repeatedly carry out reliable routine dispensation with Thermo Scientific Nalgene LDPE Dropper Bottles, made of low-density polyethylene. These bottles are to be used with Nalgene controlled dropper tips and closures.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries