Principaux documents
EP5490030001
Rotor for Eppendorf® Concentrator Plus
F-45-48-11, non aerosol tight, to hold, 48 x 1.5/2.0 mL reaction vessels
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 48 x 1.5/2.0 mL reaction vessels
fixed angle rotor (microtubes)
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490030001 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Description générale
Selection of rotors for the Eppendorf Concentrator Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG