Key Documents
Z223727
Sigma-Aldrich
Aldrich® safety bubbler
capacity 15 mL
Synonyme(s) :
bubblers, safety bubblers, scientific bubblers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Capacité
15 mL
Description générale
Built-in flash arrester bulb prevents backflow.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC