Key Documents
Z104329
Sigma-Aldrich
Aldrich® reaction vessel bubbler
joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Capacité
5-7 mL
Description générale
Excellent for closing off a reaction vessel from the atmosphere.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC