CAD7904
Animal feeding needle, non-sterile, reusable, type 304 SS
straight, size 18G, L × diam. 1.5 in. × 2.25 mm , ball
About This Item
Code UNSPSC :
42142609
Nomenclature NACRES :
NB.03
Produits recommandés
Taille
18G
Conditionnement
pkg of 12 ea
L × diam.
1.5 in. × 2.25 mm , ball
Catégories apparentées
Caractéristiques et avantages
- Fixed shape from stainless steel tubing
- Stainless steel balls
- Can be cleaned and reused as needed
- Non-sterile