Key Documents
BAH171070000
SP Bel-Art® Pipette Basket for Cleanware Pipette Rinsing System
diam. × H 10.2 cm (4 in.) × 81 cm (32 in.), 4 x 32"
Synonyme(s) :
Pipette Rinsers
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.01
Caractéristiques
to hold For 61cm (24") pipettes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H17107-0000
Diam. × H
10.2 cm (4 in.) × 81 cm (32 in.), 4 x 32"
Description générale
The Cleanware Pipette Rinsing System for pipette cleaning and storage has an effective, reliable design that ensures safe, efficient operation. Available in a complete system or in individual components.
Caractéristiques et avantages
  • Pipette Rinsers provides a simple, automatic method for rinsing large numbers of pipettes at one time
  • High level water entry provides an air gap between the maximum water level and the end of the water inlet tube ensuring safe and efficient operation
  • “Tube within a tube” siphoning system assures rapid, complete and automatic siphoning and refilling over a wide range of inlet water pressures and flow rates
  • Minimizes the danger of siphon stalling at certain water inflow rates and prevents any possible backflow into the potable water supply in the event of a pressure interruption
  • Leak-proof, polyethylene Pipette Jars store and soak pipettes and will withstand all common cleaning solutions at room temperature, including sulfuric acid/dichromate
  • Pipette Baskets, filled with pipettes, fit into the corresponding size jar; cut-out hand grips for easy handling; gusher holes provide quick drainage when lifted from a soaking jar; foam disk on the bottom cushions pipette tips to prevent breakage
  • Includes 30.5cm (12”) long piece of 2.5cm (1”) I.D. drain tube and 1.5M (5ft) of 12.5mm (½”) I.D. flexible water inlet tube
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP