Principaux documents
BAF171300018
SP Bel-Art® Pipette Jar for Cleanware Pipette Rinsing System
diam. × H 15.2 cm (6 in.) × 46 cm (18 in.)
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.01
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F17130-0018
Diam. × H
15.2 cm (6 in.) × 46 cm (18 in.)
Catégories apparentées
Description générale
Store and soak pipettes in these leakproof, molded polyethylene Pipette Jars.
Caractéristiques et avantages
  • Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP