Skip to Content
Merck
All Photos(2)

Key Documents

ABC1435M

Sigma-Aldrich

Anti-ALIX

from rabbit, purified by affinity chromatography

Synonym(s):

Programmed cell death 6-interacting protein, PDCD6-interacting protein, ALG-2-interacting protein 1, ALG-2-interacting protein X, Hp95

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PDCD6IP(10015)

General description

Programmed cell death 6-interacting protein (UniProt: Q8WUM4; also known as PDCD6-interacting protein, ALG-2-interacting protein 1, ALG-2-interacting protein X, Hp95) is encoded by the PDCD6IP (also known as AIP1, ALIX, KIAA1375) gene (gene ID: 10015) in human. ALIX, the mammalian orthologue of xenopus Xp95 and yeast BRO1, is an abundant adaptor protein involved in diverse cellular processes, including actin-based cytoskeleton assembly, integrin-mediated cell adhesion, and ESCRT- (endosomal sorting complex required for transport-) dependent membrane trafficking. ALIX is a Class E VPS protein involved in concentration and sorting of cargo proteins of the multi-vesicular body (MVB) for incorporation into intralumenal vesicles that are generated by invagination and scission from the limiting membrane of the endosome. Sorting of endocytosed membrane proteins by MVBs requires coordinated cargo recognition and assembly of membrane-sculpturing machinery that buds cargo-loaded membrane vesicles into the lumen of endosomes. MVB-mediated sorting is driven by five distinct ESCRT complexes (ESCRT-0, -I, -II and -III, and the Vps4 complex) and associated proteins. ALIX and charged MVB protein 4 (CHMP4), the mammalian orthologue of yeast Snf7, are binding partners involved in a variety of ESCRT-III-mediated membrane-remodeling processes in mammalian cells, including retroviral budding, cytokinetic abscission, biogenesis of exosomes, plasma membrane repair, and ubiquitin-independent MVB sorting of protease-activated receptor 1 (PAR1). In the absence of binding partners, an intramolecular interaction between the Patch 2 region in the Bro1 domain (aa 3-392) and the TSG101-docking site (aa 717-720) in the Pro-rich domain (aa 717-860) locks ALIX in a conformation inaccessible for interaction with CHMP4 and retroviral Gag proteins.

Specificity

This rabbit polyclonal antibody detects Programmed cell death 6-interacting protein in human and murine cells.

Immunogen

Epitope: unknown
His-tagged recombinant fragment corresponding to the first 716 amino acids of human ALIX.

Application

Anti-ALIX, Cat. No. ABC1435, is a rabbit polyclonal antibody that detects Programmed cell death 6-interacting protein and has been tested ted for use in Immunocytochemistry and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A 1:500 dilution from a representative lot detected ALIX in 10 µg of HEK293 cell lysate.

Immunocytochemistry Analysis: A representative lot detected ALIX in Immunocytochemistry applications (Morita, E., et. al. (2007). EMBO J. 26(19):4215-27).

Western Blotting Analysis: A representative lot detected ALIX in Western Blotting applications (Morita, E., et. al. (2007). EMBO J. 26(19):4215-27; Sandrin, V., et. al. (2013). Retrovirology. 10:104; Mercenne, G., et. al. (2015). Elife. 4; Zhai, Q., et. al. (2011). J Virol. 85(1):632-7).

Quality

Evaluated by Western Blotting in Hela cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ALIX in 10 µg of HeLa cell lysate.

Target description

~90 kDa observed; 96.02 kda calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service