Skip to Content
Merck
All Photos(1)

Key Documents

810180C

Avanti

TopFluor TMR PC

Avanti Research - A Croda Brand 810180C

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H82 BF2N4 O10P
CAS Number:
Molecular Weight:
1015.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810180C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810180C

concentration

1 mg/mL (810180C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Tetramethylrhodamine (TMR) phosphatidylcholine (PC) is an acyl-chain-modified fluorescently labeled phospholipid. Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes and a prime metabolite of glycerolipid metabolism.

Application

TopFluor TMR PC or 1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphocholine has been used to label biotinylated giant plasma membrane vesicles (GPMV). It is also suitable for the formation of lipid membranes in microarrays.

Biochem/physiol Actions

TopFluor-labeled lipids have spectral properties similar to tetramethylrhodamine (TMR).

Packaging

5 mL Amber Glass Screw Cap Vial (810180C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A phosphatidylinositol 4, 5-bisphosphate redistribution-based sensing mechanism initiates a phagocytosis programing.
Mu L, et al.
Nature Communications, 9(1), 4259-4259 (2018)
Single-molecule analysis of phospholipid scrambling by TMEM16F
Rikiya W, et al.
Proceedings of the National Academy of Sciences of the USA, 115(12), 3066-3071 (2018)
Rikiya Watanabe et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(12), 3066-3071 (2018-03-07)
Transmembrane protein 16F (TMEM16F) is a Ca2+-dependent phospholipid scramblase that translocates phospholipids bidirectionally between the leaflets of the plasma membrane. Phospholipid scrambling of TMEM16F causes exposure of phosphatidylserine in activated platelets to induce blood clotting and in differentiated osteoblasts to
Libing Mu et al.
Nature communications, 9(1), 4259-4259 (2018-10-17)
Phagocytosis is one of the earliest cellular functions, developing approximately 2 billion years ago. Although FcR-based phagocytic signaling is well-studied, how it originated from ancient phagocytosis is unknown. Lipid redistribution upregulates a phagocytic program recapitulating FcR-based phagocytosis with complete dependence
César Botella et al.
Progress in lipid research, 65, 12-23 (2016-11-23)
In plant cells, phosphatidylcholine (PC) is a major glycerolipid of most membranes but practically lacking from the plastid internal membranes. In chloroplasts, PC is absent from the thylakoids and the inner envelope membrane. It is however the main component of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service