Skip to Content
Merck
All Photos(1)

Key Documents

810240C

Avanti

TopFluor TMR PA

Avanti Research - A Croda Brand 810240C

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3- phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H74 BF2N4 O10P
CAS Number:
Molecular Weight:
946.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810240C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810240C

concentration

1 mg/mL (810240C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a diacyl-glycerophospholipid, which consists of a glycerol and an alcohol moiety, having two fatty acids and a phosphate group esterified at positions 1, 2, and 3, respectively.

Biochem/physiol Actions

Phosphatidic acid is a major intermediate metabolite involved in the synthesis of membrane glycerophospholipids, including phosphatidylcholine (PC), phosphatidylserine (PS), phosphatidylethanolamine (PE), phosphatidylserine (PS). PA is signaling molecule, originally synthesized in the endoplasmic reticulum (ER) and is transported to the mitochondrial outer membrane to stimulate mitofusin (Mfn)-mediated fusion.

Packaging

5 mL Amber Glass Screw Cap Vial (810240C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional Systems for Combined Delivery Biosensing and Diagnostics, 28(1), 67-76 (2020)
Brain Lipids in Synaptic Function and Neurological Disease: Clues to Innovative Therapeutic Strategies for Brain Disorders, 28(1), 67-76 (2015)
Shoichiro Kameoka et al.
Trends in cell biology, 28(1), 67-76 (2017-09-16)
Membrane organelles comprise both proteins and lipids. Remodeling of these membrane structures is controlled by interactions between specific proteins and lipids. Mitochondrial structure and function depend on regulated fusion and the division of both the outer and inner membranes. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service