Skip to Content
Merck
All Photos(1)

Key Documents

778109

Sigma-Aldrich

4,7-Dibromobenzo[c]-1,2,5-thiadiazole

99.5% (GC)

Synonym(s):

Dibromo BTD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Br2N2S
CAS Number:
Molecular Weight:
293.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.5% (GC)

form

crystalline powder

mp

184-185 °C
186-190 °C

SMILES string

Brc1ccc(Br)c2nsnc12

InChI

1S/C6H2Br2N2S/c7-3-1-2-4(8)6-5(3)9-11-10-6/h1-2H

InChI key

FEOWHLLJXAECMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This monomer is used in the synthesis of photoactive acceptor part in low band gap conjugated polymers and oligomers for high performance OPV devices

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Christopher J Takacs et al.
Journal of the American Chemical Society, 134(40), 16597-16606 (2012-09-07)
We examine the correlations of the dipole moment and conformational stability to the self-assembly and solar cell performance within a series of isomorphic, solution-processable molecules. These charge-transfer chromophores are described by a D(1)-A-D-A-D(1) structure comprising electron-rich 2-hexylbithiophene and 3,3'-di-2-ethylhexylsilylene-2,2'-bithiophene moieties
Stalder, Romain;
Chemistry of Materials, 24(16), 3143-3152 (2012)
Zi Jun Wang et al.
ChemSusChem, 8(20), 3459-3464 (2015-09-10)
Metal-free visible-light photocatalysts offer a clean, sustainable solution to many pressing environmental issues. Herein, we present a molecular design strategy to fine-tune the valence and conduction band levels of a series of conjugated microporous polymer networks based on poly(benzochalcogenadiazole) for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service