Merck
  • The status, quality, and expansion of the NIH full-length cDNA project: the Mammalian Gene Collection (MGC).
The status, quality, and expansion of the NIH full-length cDNA project: the Mammalian Gene Collection (MGC).
Genome research (2004-10-19)
Daniela S Gerhard, Lukas Wagner, Elise A Feingold, Carolyn M Shenmen, Lynette H Grouse, Greg Schuler, Steven L Klein, Susan Old, Rebekah Rasooly, Peter Good, Mark Guyer, Allison M Peck, Jeffery G Derge, David Lipman, Francis S Collins, Wonhee Jang, Steven Sherry, Mike Feolo, Leonie Misquitta, Eduardo Lee, Kirill Rotmistrovsky, Susan F Greenhut, Carl F Schaefer, Kenneth Buetow, Tom I Bonner, David Haussler, Jim Kent, Mark Kiekhaus, Terry Furey, Michael Brent, Christa Prange, Kirsten Schreiber, Nicole Shapiro, Narayan K Bhat, Ralph F Hopkins, Florence Hsie, Tom Driscoll, M Bento Soares, Tom L Casavant, Todd E Scheetz, Michael J Brown-stein, Ted B Usdin, Shiraki Toshiyuki, Piero Carninci, Yulan Piao, Dawood B Dudekula, Minoru S H Ko, Koichi Kawakami, Yutaka Suzuki, Sumio Sugano, C E Gruber, M R Smith, Blake Simmons, Troy Moore, Richard Waterman, Stephen L Johnson, Yijun Ruan, Chia Lin Wei, S Mathavan, Preethi H Gunaratne, Jiaqian Wu, Angela M Garcia, Stephen W Hulyk, Edwin Fuh, Ye Yuan, Anna Sneed, Carla Kowis, Anne Hodgson, Donna M Muzny, John McPherson, Richard A Gibbs, Jessica Fahey, Erin Helton, Mark Ketteman, Anuradha Madan, Stephanie Rodrigues, Amy Sanchez, Michelle Whiting, Anup Madari, Alice C Young, Keith D Wetherby, Steven J Granite, Peggy N Kwong, Charles P Brinkley, Russell L Pearson, Gerard G Bouffard, Robert W Blakesly, Eric D Green, Mark C Dickson, Alex C Rodriguez, Jane Grimwood, Jeremy Schmutz, Richard M Myers, Yaron S N Butterfield, Malachi Griffith, Obi L Griffith, Martin I Krzywinski, Nancy Liao, Ryan Morin, Ryan Morrin, Diana Palmquist, Anca S Petrescu, Ursula Skalska, Duane E Smailus, Jeff M Stott, Angelique Schnerch, Jacqueline E Schein, Steven J M Jones, Robert A Holt, Agnes Baross, Marco A Marra, Sandra Clifton, Kathryn A Makowski, Stephanie Bosak, Joel Malek
ABSTRACT
The National Institutes of Health's Mammalian Gene Collection (MGC) project was designed to generate and sequence a publicly accessible cDNA resource containing a complete open reading frame (ORF) for every human and mouse gene. The project initially used a random strategy to select clones from a large number of cDNA libraries from diverse tissues. Candidate clones were chosen based on 5'-EST sequences, and then fully sequenced to high accuracy and analyzed by algorithms developed for this project. Currently, more than 11,000 human and 10,000 mouse genes are represented in MGC by at least one clone with a full ORF. The random selection approach is now reaching a saturation point, and a transition to protocols targeted at the missing transcripts is now required to complete the mouse and human collections. Comparison of the sequence of the MGC clones to reference genome sequences reveals that most cDNA clones are of very high sequence quality, although it is likely that some cDNAs may carry missense variants as a consequence of experimental artifact, such as PCR, cloning, or reverse transcriptase errors. Recently, a rat cDNA component was added to the project, and ongoing frog (Xenopus) and zebrafish (Danio) cDNA projects were expanded to take advantage of the high-throughput MGC pipeline.
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, crystalline suspension in 3.8 M ammonium sulfate solution, ~350 U/mg protein (10-40 mg/ml)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized, powder, ≥4 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Inosine Monophosphate Dehydrogenase Type II human, recombinant, expressed in E. coli
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
Acyl-coenzyme A Synthetase from Pseudomonas sp., ≥2 units/mg protein
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Sigma-Aldrich
Phospholipase D from cabbage, Type IV, lyophilized powder, ≥100 units/mg solid
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Phosphorylase a from rabbit muscle, lyophilized powder, 20-30 units/mg protein
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase from rabbit muscle, Type I, ammonium sulfate suspension, 100-300 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Uridine-5′-diphosphoglucose pyrophosphorylase from baker′s yeast, Type X, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein
Sigma-Aldrich
Galactose-1-phosphate Uridyltransferase from galactose-adapted yeast, Type IV, lyophilized powder, 20-60 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein