Skip to Content
Merck
All Photos(1)

Documents

500518

Low Volume Static Mixer (50-250 μL)

housing, Binary, PEEK

Synonym(s):

ASI PEEK HOUSING, BINARY INPUT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

PEEK

fitting

10-32 ASI in-line housing for 1/16 in. O.D. tubing

compatibility

for use with ASI low volume mixer cartridge

Looking for similar products? Visit Product Comparison Guide

General description

Low Volume Mixer Housing (housing, binary) is an accessory for use with low volume mixer cartridge (50-250 μL) in ASI Static Mixers, used for HPLC and post-column derivatization. “Binary” refers to two lines going into the mixer housing. It is mainly used to combine two flowpaths into one, such as in postcolumn or gradient mixing applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service