Skip to Content
Merck
All Photos(1)

Key Documents

SML1174

Sigma-Aldrich

ML120B

≥98% (HPLC)

Synonym(s):

MLN120B, N-(6-Chloro-7-methoxy-9H-pyrido[3,4-b]indol-8-yl)-2-methyl-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£115.00

£115.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£115.00

About This Item

Empirical Formula (Hill Notation):
C19H15ClN4O2
CAS Number:
Molecular Weight:
366.80
UNSPSC Code:
12352200
NACRES:
NA.77

£115.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1cc2c([nH]c4c2ccnc4)c(c1OC)NC(=O)c3c(nccc3)C

InChI

1S/C19H15ClN4O2/c1-10-11(4-3-6-22-10)19(25)24-17-16-13(8-14(20)18(17)26-2)12-5-7-21-9-15(12)23-16/h3-9,23H,1-2H3,(H,24,25)

InChI key

ZNOLRTPMNMPLHY-UHFFFAOYSA-N

Biochem/physiol Actions

ML120B is a potent inhibitor of IKK-2 (IC50 = 60 nM). ML120B is orally bioavailable, and inhibits paw swelling, bone and cartilage degradation, and weight loss in rat adjuvant-induced arthritis.
ML120B is a potent inhibitor of IKK-2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Wang et al.
Aging, 11(18), 7402-7415 (2019-09-27)
Chemokines play a key role in post-traumatic inflammation and secondary injury after spinal cord injury (SCI). CCL28, the chemokine CC-chemokine ligand 28, is involved in the epithelial and mucosal immunity. However, whether CCL28 participates in the physiopathologic processes after SCI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service