Skip to Content
Merck
All Photos(1)

Documents

40005

Supelco

Benzidine solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2

InChI key

HFACYLZERDEVSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoheb Karim et al.
Environmental technology, 32(1-2), 83-91 (2011-04-09)
Bitter gourd peroxidase entrapped on calcium-alginate-starch beads was employed for the treatment of water, polluted with benzidine, in a batch process as well as in a continuous reactor. The immobilized enzyme had the same pH and temperature optima as the
Jungsun Park et al.
American journal of industrial medicine, 52(8), 625-632 (2009-06-30)
Many corporations move their manufacturing facilities or technologies from developed to developing countries. Stringent regulations have made it costly for industries to operate in developed, industrialized countries. In addition, labor costs are high in these countries, and there is increasing
Occupational contact allergy caused by benzidine in three tannery workers.
Sri Awalia Febriana et al.
Contact dermatitis, 66(6), 345-346 (2012-05-10)
Rosa I Gallagher et al.
Methods in molecular biology (Clifton, N.J.), 723, 275-301 (2011-03-04)
The Reverse Phase Protein Microarray (RPMA) is an array platform used to quantitate proteins and their posttranslationally modified forms. RPMAs are applicable for profiling key cellular signaling pathways and protein networks, allowing direct comparison of the activation state of proteins
Jung Woo Lee et al.
Chemical communications (Cambridge, England), 48(3), 422-424 (2011-11-15)
We report a facile route to synthesize size tunable Fe(3)O(4) nanoparticles (NPs)-carbon nitride nanotube (CNNT) hybrids. These hybrids showing the water-soluble property are proven to exhibit ultra high peroxidase mimetic activity compared to those of pure NPs, where a colorless

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service