Merck
  • Reverse phase protein microarrays: fluorometric and colorimetric detection.
Reverse phase protein microarrays: fluorometric and colorimetric detection.
Methods in molecular biology (Clifton, N.J.) (2011-03-04)
Rosa I Gallagher, Alessandra Silvestri, Emanuel F Petricoin, Lance A Liotta, Virginia Espina
ABSTRACT
The Reverse Phase Protein Microarray (RPMA) is an array platform used to quantitate proteins and their posttranslationally modified forms. RPMAs are applicable for profiling key cellular signaling pathways and protein networks, allowing direct comparison of the activation state of proteins from multiple samples within the same array. The RPMA format consists of proteins immobilized directly on a nitrocellulose substratum. The analyte is subsequently probed with a primary antibody and a series of reagents for signal amplification and detection. Due to the diversity, low concentration, and large dynamic range of protein analytes, RPMAs require stringent signal amplification methods, high quality image acquisition, and software capable of precisely analyzing spot intensities on an array. Microarray detection strategies can be either fluorescent or colorimetric. The choice of a detection system depends on (a) the expected analyte concentration, (b) type of microarray imaging system, and (c) type of sample. The focus of this chapter is to describe RPMA detection and imaging using fluorescent and colorimetric (diaminobenzidine (DAB)) methods.
Product Description
Supelco
Benzidine solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Benzidine, ≥98.0% (N)
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)
Supelco
Benzidine, analytical standard