Skip to Content
Merck
All Photos(1)

Key Documents

D002

Sigma-Aldrich

(±)-2-Amino-6,7-dihydroxy-1,2,3,4-tetrahydronaphthalene hydrobromide

solid

Synonym(s):

6,7-ADTN hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2 · HBr
CAS Number:
Molecular Weight:
260.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage condition

protect from light

solubility

H2O: >10 mg/mL (Dissolve in oxygen-free water containing 0.1% sodium metabisulfite or other antioxidants.)
aqueous base: unstable

storage temp.

2-8°C

SMILES string

Br[H].NC1CCc2cc(O)c(O)cc2C1

Biochem/physiol Actions

Dopamine receptor agonist.

Caution

Air- and light-sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Balmforth et al.
Journal of neurochemistry, 51(5), 1510-1515 (1988-11-01)
3,4-Dihydroxyphenylethylamine (dopamine) and beta-adrenergic receptor agonists and antagonists were assessed for their effects on cyclic AMP accumulation in human astrocytoma derived clone D384 cells. Dopamine, SKF 38393, and 2-amino-6,7-dihydroxy-1,2,3,4-tetrahydronaphthalene increased cyclic AMP content with Ka values of 2.0, 0.2, and
S Patel et al.
Behavioral neuroscience, 102(4), 596-600 (1988-08-01)
The effects of administration of gamma-aminobutyric acid (GABA) compounds into the ventral pallidum and substantia innominata on the locomotor hyperactivity induced by the dopamine agonist 2-amino-6,7-dihydroxy-1,2,3,4-tetrahydronaphthalene (ADTN) in the nucleus accumbens were investigated in rats. Hyperactivity induced by ADTN was
U Sreenivasan et al.
Journal of medicinal chemistry, 36(2), 256-263 (1993-01-22)
A series of analogues of the potent analogue of Pro-Leu-Gly-NH2 (PLG), 2-oxo-3(R)-[(2(S)-pyrrolidinylcarbonyl)amino]-1-pyrrolidineacet amide (2) were synthesized in which the (R)-gamma-lactam residue of 2 was replaced with a (R)-beta-lactam, (R)-aminosuccinimide, (R)-cycloseryl, (R)-delta-lactam, (R)-epsilon-lactam, or (S)-epsilon-lactam residue to give analogues 3-8, respectively.
S B Ross
Journal of neurochemistry, 56(1), 22-29 (1991-01-01)
The concentration of dopamine (DA) in the synaptic cleft in the mouse striatum in vivo was estimated from the competition between the synaptic DA and the 3H-labelled DA D2 receptor agonists N-n-propylnorapomorphine (NPA) or N,N-diethyl-N'-[(3 alpha, 4a alpha, 10 beta)-1,2,3,4,4a,5,10,10a-octahydro-
G M Ross et al.
Journal of molecular neuroscience : MN, 4(3), 141-148 (1993-01-01)
Several pharmacologically active catecholamines have been shown to react covalently with CNS proteins, namely species of 47, 40, 22, and 20 kDa. Of these, the 47-kDa protein showed the greatest incorporation of tritium following treatment with [3H]dopamine, [3H]ADTN, or [3H]N-propyl-norapomorphine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service