Merck
  • Synaptic concentration of dopamine in the mouse striatum in relationship to the kinetic properties of the dopamine receptors and uptake mechanism.
Synaptic concentration of dopamine in the mouse striatum in relationship to the kinetic properties of the dopamine receptors and uptake mechanism.
Journal of neurochemistry (1991-01-01)
S B Ross
ABSTRACT
The concentration of dopamine (DA) in the synaptic cleft in the mouse striatum in vivo was estimated from the competition between the synaptic DA and the 3H-labelled DA D2 receptor agonists N-n-propylnorapomorphine (NPA) or N,N-diethyl-N'-[(3 alpha, 4a alpha, 10 beta)-1,2,3,4,4a,5,10,10a-octahydro- 7-hydroxyl-1-propyl-3-benzo (g) quinolinyl]sulfamide (Sandoz 205-501) injected intravenously in tracer doses. Knowing the inhibitor constant for DA in inhibiting the binding of these receptor agonists in vitro, attempts were made to calculate the changes in the synaptic DA concentration from the changes in the in vivo binding of the receptor agonists evoked by various pharmacological agents. Inhibiting the firing of the dopaminergic neurons by gamma-butyrolactone (GBL) increased the binding of the receptor agonists corresponding to a decrease in the synaptic DA concentration of 55 +/- 2 nM in the experiments with [3H]Sandoz 205-501 and 48 +/- 3 nM in the experiments with tracer doses of [3H]NPA. These values may therefore approximate the normal DA concentration in the synaptic cleft in the mouse striatum. With this technique it was also possible to determine the synaptic concentration of NPA by its competition with [3H]Sandoz 205-501 for the DA D2 receptors in the striatum of GBL-treated mice in vivo. To compare the estimated synaptic concentration of DA with the affinity of DA to D1 and D2 receptors and to the DA transporter in the mouse striatum the kinetic parameters were determined at 37 degrees C in vitro.(ABSTRACT TRUNCATED AT 250 WORDS)