Skip to Content
Merck
All Photos(1)

Documents

311

90070547, insect embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

311, 90070547

biological source

insect embryo

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Drosophila embryo

Cell Line Description

Derived from embryonic cells of Drosophila melanogaster. Useful for virus studies, particularly picornaviruses and rhabdoviruses.

Application

Virus studies: picornaviruses, rhabdoviruses

Culture Medium

Schneider′s Drosophila medium + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. Cells may semi-adhere to the tissue culture flask, shake gently to dislodge; culture at 25°C; No CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service