A2B5 clone 105
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-Retinal Neuron Glycolipid, chick, 90070501
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 90070501-DNA-5UG
pkg of vial of cells 90070501-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
P3X63Ag8
Reactivity
Anti-Retinal Neuron Glycolipid, chick
technique(s)
cell culture | mammalian: suitable
isotype
IgM
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3X63Ag8 x Mouse BALB/c spleen
Cell Line Description
This cytotoxic antibody is directed against a glycolipoid present on chick embryo retinal neuron cell bodies. The antigen is found in chick embryo brain, spinal cord and dorsal root ganglia neurons and in bovine and human brain. The antigen is not present on axons or dendrites.
Immunogen
Chicken embryo retina cells
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service