Skip to Content
Merck
All Photos(1)

Key Documents

BHK21C13-3P

84100501, hamster kidney, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

BHK21C13-3P, 84100501

biological source

hamster kidney

growth mode

Adherent

karyotype

2n = 44, pseudodiploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Syrian kidney

Cell Line Description

BHK21C13-3P is a subclone of the parent line BHK-21 (Clone-13) (Sigma Catalogue number. 85011433). The cells grow as monolayers and have the same virus susceptibility as the parent line.

Application

Virus replication studies

Culture Medium

GMEM + 2mM Glutamine + 5% Tryptose Phosphate Broth + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service