Recommended Products
product name
BHK21C13-2P, 84111301
biological source
hamster kidney
growth mode
Suspension
karyotype
2n = 44, pseudodiploid
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Hamster Syrian kidney
Cell Line Description
Derived from the parent line BHK-21 (Clone-13) (Sigma Catalogue number. 85011433). The cells have been adapted to growth in suspension and have the same virus susceptibility as the parent line.
Application
Virus replication studies
Culture Medium
GMEM + 2mM Glutamine + 5% Tryptose Phosphate Broth + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 2-9x100,000 cells/ml (split ratio of 1:3 to 1:6); 5% CO2; 37°C. Cells need to be grown in a techni-stirrer or they may start to re-attach.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service