Skip to Content
Merck
All Photos(1)

Key Documents

37201

Supelco

Sulfite test sticks Quantofix®

10-1000 mg/L (SO3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

sulfite

measuring range

10-1000 mg/L (SO3)

application(s)

environmental
general analytical

detection method

colorimetric

General description

Contents:
Tube with 100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).

Gradation:
0 - 10 - 25 - 50 - 100 - 250 - 500 - 1000 mg/l SO32-

Application

It was used in characterizing Na233SO3, in an attempt to demonstrate the presence of coordinated sulfate, in the reaction pathway of Arabidopsis thaliana sulfite oxidase, using 33S labeling and electron spin echo envelope modulation spectroscopy.

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrei V Astashkin et al.
Journal of the American Chemical Society, 129(47), 14800-14810 (2007-11-07)
Sulfite oxidase from Arabidopsis thaliana has been reduced at pH = 6 with sulfite labeled with 33S (nuclear spin I = 3/2), followed by reoxidation by ferricyanide to generate the Mo(V) state of the active center. To obtain information about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service