Skip to Content
Merck
All Photos(1)

Documents

35405

Supelco

α-Zearalanol solution

~10 μg/mL in acetonitrile, analytical standard

Synonym(s):

(3S,7R)-3,4,5,6,7,8,9,10,11,12-Decahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one solution, 2,4-Dihydroxy-6-(6α,10-dihydroxyundecyl)benzoic acid μ-lactone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

not available in Canada

concentration

~10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@H](O)CCCCCc2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14+/m0/s1

InChI key

DWTTZBARDOXEAM-GXTWGEPZSA-N

General description

α-Zearalanol is a resorcyclic acid lactone. It is a plant derived, nonsteroidal phytoestrogen, often used as a dietary supplement.

Application

α-Zearalanol may be used as an analytical reference standard for the determination of the analyte in traditional Chinese medicines, human and animal urine by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid method for simultaneous determination of zearalenone, α-zearalenol, β-zearalenol, zearalanone, α-zearalanol and β-zearalanol in traditional Chinese medicines by ultra-high-performance liquid chromatography-tandem mass spectrometry.
Han Z, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879(5-6), 411-420 (2011)
Rui Feng et al.
Biosensors & bioelectronics, 42, 367-372 (2012-12-06)
Nanoporous PtCo alloy was designed as an antibody carrier for preparation of a highly sensitive immunosensor. The immunosensor was constructed by assembling the capture zeranol antibody on thionine decorated graphene nanosheets modified glassy carbon electrode. With an enzyme-free immunosensor mode
Nidia V Valenzuela-Grijalva et al.
Journal of the science of food and agriculture, 92(7), 1362-1367 (2011-12-07)
The effect of zeranol implantation strategy on intramuscular fat, fatty acid profile and cholesterol content of the longissimus dorsi muscle of hair lambs was studied. Four treatments were tested: C, control group; Z12, 12 mg zeranol; Z24, 24 mg zeranol
Hui Zhou et al.
Nature chemical biology, 8(4), 331-333 (2012-03-13)
Iterative highly reducing polyketide synthases from filamentous fungi are the most complex and enigmatic type of polyketide synthase discovered to date. Here we uncover an unusual degree of programming by the hypothemycin highly reducing polyketide synthase, in which a single
Yuan Gao et al.
The Analyst, 137(1), 229-236 (2011-11-04)
Although anti-zearalenone (ZEN) antibodies have been widely prepared, these antibodies cross-react with α-zearalenol (α-ZOL), β-zearalenol (β-ZOL), zearalanone (ZAN), α-zearalanol (α-ZAL) and β-zearalanol (β-ZAL). To overcome this problem and improve the specificity of immunoassays, we produced anti-ZEN antibodies based on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service