Merck
  • Preparation of highly specific anti-zearalenone antibodies by using the cationic protein conjugate and development of an indirect competitive enzyme-linked immunosorbent assay.
Preparation of highly specific anti-zearalenone antibodies by using the cationic protein conjugate and development of an indirect competitive enzyme-linked immunosorbent assay.
The Analyst (2011-11-04)
Yuan Gao, Meihua Yang, Cheng Peng, Xiaohong Li, Runlan Cai, Yun Qi
ABSTRACT
Although anti-zearalenone (ZEN) antibodies have been widely prepared, these antibodies cross-react with α-zearalenol (α-ZOL), β-zearalenol (β-ZOL), zearalanone (ZAN), α-zearalanol (α-ZAL) and β-zearalanol (β-ZAL). To overcome this problem and improve the specificity of immunoassays, we produced anti-ZEN antibodies based on a ZEN-cationic protein conjugate. In this study, ZEN was coupled with cationic bovine serum albumin (cBSA) via a Mannich reaction. After BALB/c mice were immunized with ZEN-cBSA, an immunological response was rapidly induced. The titers of the polyclonal antisera and monoclonal antibody were 30,000 and 20,000, respectively. Cross-reactivity (CR) values of the anti-ZEN polyclonal antisera and monoclonal antibody with the 5 analogs were <7% and <2%, respectively. An indirect competitive enzyme-linked immunosorbent assay based on the monoclonal anti-ZEN antibody was established. The recovery rates of ZEN in spiked cereal and feed were in the range of 80%-120% with coefficients of variation <15%. The intra-assay variation and inter-assay variation in assay buffer were both <5%. Therefore, the results demonstrated a feasible approach for preparing highly specific, higher titer and more rapidly induced antibodies against ZEN by using a ZEN-cBSA conjugate as the immunogen instead of currently used immunogens.
Sigma-Aldrich
α-Zearalanol, ~98% (HPLC)