Skip to Content
Merck

1.06400

SAFC

Sodium chloride

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
£28.00
5 KG
£45.00
25 KG
£144.00
50 KG
£259.00
100 KG
£458.00
25 KG
£560.00
100 KG
£819.00
500 KG
Inquire

£28.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
£28.00
5 KG
£45.00
25 KG
£144.00
50 KG
£259.00
100 KG
£458.00
25 KG
£560.00
100 KG
£819.00
500 KG
Inquire

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-598-3

£28.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

BP
Ph. Eur.
USP

Quality Level

vapor pressure

1.3 hPa ( 865 °C)

product line

EMPROVE® ESSENTIAL

form

solid

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

solubility

358 g/L

density

2.17 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical

storage temp.

2-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium chloride is a mineral salt with broad range of applications in pharmaceutical formulations. It is typically used as an osmotic agent in liquid dosage forms e.g. for preparation of isotonic formulations, as bulking or coating agent for solid dosage forms, as well as for semi-solid dosage forms like creams.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service