Skip to Content
Merck
All Photos(1)

Key Documents

567442

Millipore

Sodium Chloride Tablets

Each tablet yields 1 L of 0.9% sodium chloride.

Synonym(s):

Sodium chloride, NaCl 10 Tablets, Halite, NaCl

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£56.40

£56.40


Estimated to ship on31 March 2026Details


Request a Bulk Order

Select a Size

Change View
1 EA
£56.40

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25
grade:
for analytical purposes

£56.40


Estimated to ship on31 March 2026Details


Request a Bulk Order

grade

for analytical purposes

Quality Level

packaging

pkg of 10 tablets

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

mp

801 °C (lit.)

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service