Skip to Content
Merck
All Photos(1)

Key Documents

W360007

Sigma-Aldrich

4-(Methylthio)butanol

97%

Synonym(s):

4-(Methylthio)-1-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3S(CH2)4OH
CAS Number:
Molecular Weight:
120.21
FEMA Number:
3600
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.078
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

Assay

97%

refractive index

n20/D 1.487 (lit.)
n20/D 1.487

bp

95-98 °C/20 mmHg (lit.)
95-98 °C/20 mmHg

density

0.993 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cabbage; sulfurous

SMILES string

CSCCCCO

InChI

1S/C5H12OS/c1-7-5-3-2-4-6/h6H,2-5H2,1H3

InChI key

JNTVUHZXIJFHAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service