Skip to Content
Merck
All Photos(1)

Documents

W316202

Sigma-Aldrich

Furfuryl thioacetate

≥98%, FG

Synonym(s):

S-Furfuryl thioacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20
FEMA Number:
3162
EC Number:
Council of Europe no.:
2250
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.033
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥98%

refractive index

n20/D 1.526 (lit.)

bp

90-92 °C/12 mmHg (lit.)

density

1.171 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

burnt; coffee; roasted; sulfurous

SMILES string

CC(SCC1=CC=CO1)=O

InChI

1S/C7H8O2S/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

LQOUTUIIYXYBQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Furfuryl thioacetate is a sulfur-containing flavor compound mainly found in coffee.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New developments in coffee aroma research.
Flament Y.
Journal of Agricultural and Food Chemistry, 15(1), 15-23 (1967)
Precursors of sulfur?containing flavor compounds
Schutte L & Teranishi R
Critical Reviews in Food Science and Nutrition, 4(4), 457-505 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service