N6265
2-(5-Nitro-2-pyridylazo)-5-(N-propyl-N-sulfopropylamino)phenol disodium salt
≥95%
Synonym(s):
Nitro-PAPS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C17H19N5O6SNa2
CAS Number:
Molecular Weight:
467.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥95%
storage temp.
2-8°C
SMILES string
[Na+].[Na+].CCCN(CCCS([O-])(=O)=O)c1ccc(\N=N\c2cc(ccn2)[N+]([O-])=O)c([O-])c1
InChI
1S/C17H21N5O6S.2Na/c1-2-8-21(9-3-10-29(26,27)28)13-4-6-15(16(23)11-13)19-20-17-7-5-14(12-18-17)22(24)25;;/h4-7,11-12,23H,2-3,8-10H2,1H3,(H,26,27,28);;/q;2*+1/p-2/b20-19+;;
InChI key
YGNYDGHRNNUMAZ-LLIZZRELSA-L
Application
Reagent for photometric determination of iron.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Makino, T., et al.
Proc. 26th Ann. Mtg. Japan Soc. Clin. Chem, 107-107 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service