Skip to Content
Merck
All Photos(1)

Key Documents

803596

Sigma-Aldrich

BM(PEG)3 (1,11-bismaleimido-triethyleneglycol)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O7
Molecular Weight:
352.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

form

powder

mol wt

352.34

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C=CC1=O)N1CCOCCOCCOCCN2C(C=CC2=O)=O

InChI

1S/C16H20N2O7/c19-13-1-2-14(20)17(13)5-7-23-9-11-25-12-10-24-8-6-18-15(21)3-4-16(18)22/h1-4H,5-12H2

InChI key

OYRSKXCXEFLTEY-UHFFFAOYSA-N

General description

BM(PEG)2 and BM(PEG)3 are homobifunctional, maleimide crosslinkers for conjugation between sulfhydryl groups (-SH). Such bismaleimide crosslinkers are commonly used to explore and characterize protein structure (i.e. oligomerization) or protein interactions. Because BM(PEG)2 and BM(PEG)3 have the same reactivity but differ in length, the relative success of these and similar reagents in forming crosslinks between sites in a protein oligomer or interaction can assist in determining intra- and intermolecular distances.

Features and Benefits

  • Reactive groups: maleimide (both ends)
  • Reactive towards: sulfhydryl groups
  • Long, pegylated, sulfhydryl-to-sulfhydryl crosslinkers, composed of maleimide groups and 3-unit polyethylene glycol spacer arm
  • PEG spacers provide unique advantages, including enhanced solubility, increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Pure compounds with defined structure and molecular weight, ensuring reproducible protein-modification effects
  • Ideal for small molecule or peptide conjugations

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Jastrzebska et al.
The Journal of biological chemistry, 279(52), 54663-54675 (2004-10-19)
Rhodopsin (Rho) is a G protein-coupled receptor that initiates phototransduction in rod photoreceptors. High expression levels of Rho in the disc membranes of rod outer segments and the propensity of Rho to form higher oligomeric structures are evident from atomic
Regina B Troyanovsky et al.
Molecular and cellular biology, 23(22), 7965-7972 (2003-10-31)
E-cadherin is a transmembrane protein that mediates Ca(2+)-dependent cell-cell adhesion. To study cadherin-cadherin interactions that may underlie the adhesive process, a recombinant E-cadherin lacking free sulfhydryl groups and its mutants with novel cysteines were expressed in epithelial A-431 cells. These
Kyoko Takebe et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(14), 2037-2047 (2003-11-05)
Epimorphin is a mesenchymal morphogen that has been shown to mediate epithelial-mesenchymal signaling interactions in various organs. We now show that epimorphin functions in hair follicle morphogenesis; using a novel ex vivo organ culture assay, we define a mechanism for
Ching-Jou Lim et al.
Pharmaceutical research, 21(11), 1985-1992 (2004-12-14)
To investigate if the cross-linking of transferrin receptor (TfR) induced by Tf-oligomers alters the endocytosis of receptor-ligand complexes in cultured tumor cells and hence increases intracellular drug release. An average of 3.5 Tf molecules per aggregate were cross-linked either by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service