Merck
  • Transferrin-oligomers as potential carriers in anticancer drug delivery.
Transferrin-oligomers as potential carriers in anticancer drug delivery.
Pharmaceutical research (2004-12-14)
Ching-Jou Lim, Wei-Chiang Shen
ABSTRACT
To investigate if the cross-linking of transferrin receptor (TfR) induced by Tf-oligomers alters the endocytosis of receptor-ligand complexes in cultured tumor cells and hence increases intracellular drug release. An average of 3.5 Tf molecules per aggregate were cross-linked either by using homobifunctional linker (1, 11-bis-maleimidotetraethyleneglycol) [Tf(3.5-BM(PEO)4)] or heterobifunction linker [succinimidyl 4-(-p-maleimidophenyl)-butyrate] (Tf(3.5-SMPB)). Cell surface binding and competition experiments with 125I-Tf for TfR binding were studied to demonstrate that Tf-oligomers maintain specificity of the TfR-binding. To determine the degradation of Tf-oligomers in TfR-mediated endocytosis, cultured tumor cells were pulsed for 15 min with 125I-Tf-oligomers and chased for 2 h at 37 degrees C in the presence of excess unlabeled Tf. The chase medium was subjected to TCA precipitation to separate the intact and degraded Tf. To investigate if the alteration of TfR-trafficking facilitates the intracellular release of the drug from the Tf-conjugated form, methotrexate (MTX) was conjugated to Tf-oligomer (Agg-Tf-MTX) and its antiproliferative activity was compared with monomeric-Tf-MTX (Mono-Tf-MTX) in human colon carcinoma (Caco-2) cells, human breast adenocarcinoma (MCF-7) cells, wild-type Chinese hamster ovary (CHO) cells, and MTX-resistant CHO (CHO-MTX-RII) cells. TfR-mediated degradation of Tf-oligomers was higher than that of monomeric Tf in both Caco-2 and MCF-7 cells. The IC50 of Agg-Tf-MTX was lower than that of Mono-Tf-MTX in both tumor cell lines. The IC50 of MTX and Mono-Tf-MTX in CHO-MTX-RII cells was higher than that in wild-type CHO cells, whereas the Agg-Tf-MTX was almost identical in both the resistant and wild-type cells. Cross-linking of TfR induced by oligomeric Tf binding alters the intracellular trafficking of Tf-TfR complexes, redirects them out of the recycling pathway, and targets them to intracellular degradation in cultured tumor cells. The alteration of TfR-trafficking facilitates the intracellular release of the drug from the Tf-conjugated form. Consequently, Agg-Tf-MTX is more effective than Mono-Tf-MTX as a TfR-mediated antiproliferative agent in tumor cells, as well as in MTX-resistant transport deficient cells. Therefore, Tf-oligomers are potentially effective TfR-targeting carriers for intracellular delivery of anticancer drugs.
Sigma-Aldrich
BM(PEG)2 (1,8-bismaleimido-diethyleneglycol)