Skip to Content
Merck
All Photos(1)

Key Documents

129720

Sigma-Aldrich

4-(Methylamino)phenol hemisulfate salt

98%

Synonym(s):

Metol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H9NO · 1/2H2SO4
CAS Number:
Molecular Weight:
172.19
Beilstein:
3919382
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

Assay

98%

autoignition temp.

989 °F

mp

260 °C (dec.) (lit.)

SMILES string

OS(O)(=O)=O.CNc1ccc(O)cc1.CNc2ccc(O)cc2

InChI

1S/2C7H9NO.H2O4S/c2*1-8-6-2-4-7(9)5-3-6;1-5(2,3)4/h2*2-5,8-9H,1H3;(H2,1,2,3,4)

InChI key

ZVNPWFOVUDMGRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service