Skip to Content
Merck
All Photos(1)

Key Documents

320013

Sigma-Aldrich

4-(Methylamino)phenol hemisulfate salt

ACS reagent, 99%

Synonym(s):

p-(Methylamino)phenol hemisulfate salt, Metol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
£31.20
500 G
£127.00

£31.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
£31.20
500 G
£127.00

About This Item

Linear Formula:
C7H9NO · 1/2H2SO4
CAS Number:
Molecular Weight:
172.19
Beilstein:
3919382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

£31.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent

Assay

99%
99.0-101.5% (ACS specification)

form

powder

autoignition temp.

989 °F

ign. residue

≤0.1%

mp

260 °C (dec.) (lit.)

suitability

passes test for determination of phosphate

functional group

amine

SMILES string

OS(O)(=O)=O.CNc1ccc(O)cc1.CNc2ccc(O)cc2

InChI

1S/2C7H9NO.H2O4S/c2*1-8-6-2-4-7(9)5-3-6;1-5(2,3)4/h2*2-5,8-9H,1H3;(H2,1,2,3,4)

InChI key

ZVNPWFOVUDMGRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-(Methylamino)phenol hemisulfate salt can be used in the synthesis of 4[N-methyl-N(4-hydroxyphenyl)amino]-7-nitrobenzofurazan (NBFhd), a novel fluorescent sensor for reactive oxygen species.[1]

Legal Information

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

494.6 °F - closed cup

Flash Point(C)

257 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of a new fluorescent probe for reactive oxygen species.
Heyne B
Organic & Biomolecular Chemistry, 5(9), 1454-1458 (2007)
Ayman A Gouda et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 130, 480-487 (2014-05-13)
Simple, rapid and accurate high performance liquid chromatographic (HPLC) and spectrophotometric methods are described for determination of antihistaminic acrivastine in capsules. The first method (method A) is based on accurate, sensitive and stability indicating chromatographic separation method. Chromolith® Performance RP-18e

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service