Principaux documents
Z744004
Worthington Accessories
rack for 5 drawers for LS3000 liquid nitrogen refridgerator
1 EA
275,00 €
275,00 €
Check Cart for Availability
Changer de vue
1 EA
275,00 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
275,00 €
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Worthington RS30-9C50
Technique(s)
cryopreservation: suitable
Catégories apparentées
Description générale
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.