Principaux documents
Z742396
PELCO® Sliding Latch Microslide Storage Boxes
black, Holds 100 x slides 76x26 mm
Synonyme(s) :
slide storage
12 EA
210,00 €
210,00 €
Check Cart for Availability
Changer de vue
12 EA
210,00 €
About This Item
Code UNSPSC :
41123302
Nomenclature NACRES :
MA.01
210,00 €
Check Cart for Availability
Matériaux
acrylonitrile-butadiene-styrene (ABS)
black
Caractéristiques
Holds 100 x slides 76x26 mm
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Ted Pella 2302-6-CS
L × l × H
209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.)
Description générale
Exceptional slide storage containers providing maximum protection for your microscope slides.
Caractéristiques et avantages
  • Numbered index sheet in lid for quick identification
  • Foam liner
  • Stackable
  • ABS-durable/impact resistant
  • Hinged lid
  • Unique sliding compression latch
Informations légales
PELCO is a registered trademark of Ted Pella, Inc