Principaux documents
TMO312085-0032
Nalgene® narrow-mouth bottle
amber high-density polyethylene, volume 1000 mL, case of 50 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
50 EACH
179,00 €
179,00 €
Date d'expédition estimée le09 avril 2025
Changer de vue
50 EACH
179,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
179,00 €
Date d'expédition estimée le09 avril 2025
Matériaux
amber high-density polyethylene
Conditionnement
case of 50 ea
Fabricant/nom de marque
Nalgene 312085-0032
Capacité
1000 mL
Volume
1000 mL
Description générale
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries