Principaux documents
TMO161093
Nunc® MicroWell 96-well Plate with high flange skirt
Nunclon® Delta surface, polystyrene, sterile, with lid, clear, case of 160 ea
Synonyme(s) :
multiwell plate, treated multiwell plate, treated plate
160 EACH
1 090,00 €
1 090,00 €
Check Cart for Availability
Changer de vue
160 EACH
1 090,00 €
About This Item
Code UNSPSC :
41122102
Nomenclature NACRES :
NB.15
1 090,00 €
Check Cart for Availability
Matériaux
clear
polystyrene
Stérilité
sterile
Conditionnement
case of 160 ea
Fabricant/nom de marque
Nunc 161093
Catégories apparentées
Description générale
Obtain consistent and reliable results with Thermo Scientific Nunc MicroWell 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries