Principaux documents
T4418
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 38 mL
Synonyme(s) :
High-Speed ​​Centrifuge Tubes, Nalgene tubes
10 EA
84,00 €
84,00 €
Check Cart for Availability
Changer de vue
10 EA
84,00 €
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.11
84,00 €
Check Cart for Availability
Matériaux
polycarbonate
unlipped
Stérilité
non-sterile
Caractéristiques
RCF 50,000 × g
Conditionnement
pack of 10
Fabricant/nom de marque
Nalgene® 3117-0380
Capacité nominale
38 mL
Description générale
These high-speed polycarbonate tubes withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Designed with round bottoms to fit high-speed rotors
  • Molding technique produces uniform wall thickness for increased strength
  • Uniform diameter (no taper) for better rotor fit and force tolerance
  • Lip allows easier retrieval from rotor cavities
  • Economical choice - may also be used as test tubes
  • Longer-lasting than polycarbonate tubes under conditions of repeated autoclaving
  • Autoclavable/Transparent
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries