Accéder au contenu
Merck
Toutes les photos(1)

Principaux documents

GE17-5175-01

Superdex® 200 10/300 GL

Cytiva 17-5175-01, column L × I.D. 30 cm × 10 mm, 13 μm avg. part. size

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Code UNSPSC :
12352200

Matériaux

borosilicate glass column
polypropylene filter

Poids mol.

10000-600000 Da (globular proteins)

Conditionnement

pkg of 1 ea

Fabricant/nom de marque

Cytiva 17-5175-01

Paramètres

0.25-0.75 mL/min flow rate
1.5 Mpa (217 psi)
25-500 mL sample volume

Technique(s)

HPLC: suitable

Volume du lit

24 mL

Colonne, L × D.I.

30 cm × 10 mm

Matrice

agarose-dextran composite

Taille moy. des particules

13 μm

cleaning

1-14

working range

3-12

Technique de séparation

size exclusion (SEC)

Description générale

Superdex® 200 10/300 GL is a prepacked gel filtration column for high-resolution, semipreperative and analytical separations of biomolecules. Superdex® 200 has a separation range for molecules with molecular weights between 10 000 and 600 000.

Superdex® prepacked columns are designed for high-performance, laboratory-scale separations of proteins, peptides, and other biomolecules according to size. High performance gel filtration offers fast separations with high resolution for a variety of applications including protein purification, studies of complex formation, and screening of uncharacterized samples.

Superdex® 75 5/150 GL and Superdex® 200 5/150 GL have small bed volumes, which is useful for a range of applications such as screening solubilization conditions for membrane proteins, the study of protein-protein interactions, and rapid purity check of proteins. These columns are an excellent choice when time and sample/buffer consumption is more important than the highest resolution since less buffer and sample volumes are required.

Superdex® columns 10/300 GL and 5/150 GL are prepacked Tricorn columns, see Tricorn Column Family, that provideexcellent reproducibility and durability in purification using a chromatography system.

Caractéristiques et avantages

  • High-resolution separations of proteins, peptides, and other biomolecules according to size.
  • Highly suitable for the polishing step in a purification procedure.
  • Easy and predictable scale-up.
  • Excellent reproducibility and durability.
  • For rapid protein homogeneity analysis or purity check use the 5/150 GL short columns.

Adéquation

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 30% isopropanol, 70% ethanol, 1 M NaOH (for cleaning in place)
for high-resolution, semipreperative and analytical separations of biomolecules

Stockage et stabilité

Store at 4 to 30 °C (20% Ethanol)

Remarque sur l'analyse

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Informations légales

Superdex is a registered trademark of Cytiva

Pictogrammes

Flame

Mention d'avertissement

Warning

Mentions de danger

Code de la classe de stockage

"" - ""


Certificats d'analyse (COA)

Recherchez un Certificats d'analyse (COA) en saisissant le numéro de lot du produit. Les numéros de lot figurent sur l'étiquette du produit après les mots "Lot" ou "Batch".

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique