Principaux documents
GBL505170
Grace Bio-Labs ONCYTE® NOVA nitrocellulose film slides
1- 20 mm x 51 mm NC pad per slide, frosted end glass
Synonyme(s) :
nitrocellulose slides, protein microarrays
20 EA
637,00 €
637,00 €
Check Cart for Availability
Changer de vue
20 EA
637,00 €
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.15
637,00 €
Check Cart for Availability
Matériaux
nitrocellulose (on glass)
Conditionnement
pack of 20 ea
Fabricant/nom de marque
Grace Bio-Labs 505170
Taille externe
25 mm × 75 mm
Type de liaison
medium binding surface
Description générale
Grace Bio-Labs ONCYTE® NOVA nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Informations légales
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.