Accéder au contenu
Merck
Toutes les photos(1)

Principaux documents

54804

Sigma-Aldrich

1-Hydroxybenzotriazole hydrate

puriss., ≥99.0% (HPLC)

Synonyme(s) :

HOBt Hydrate

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Formule empirique (notation de Hill):
C6H5N3O · xH2O
Numéro CAS:
Poids moléculaire :
135.12 (anhydrous basis)
Beilstein:
4515
Numéro CE :
Code UNSPSC :
12352000
ID de substance PubChem :

Qualité

puriss.

Essai

≥99.0% (HPLC)

Impuretés

11-13% water

Pf

155-158 °C (lit.)

Solubilité

DMF: 100 mg/mL, clear

Chaîne SMILES 

[H]O[H].On1nnc2ccccc12

InChI

1S/C6H5N3O.H2O/c10-9-6-4-2-1-3-5(6)7-8-9;/h1-4,10H;1H2

Clé InChI

PJUPKRYGDFTMTM-UHFFFAOYSA-N

Vous recherchez des produits similaires ? Visite Guide de comparaison des produits

Remplacé(e)(s) par

Réf. du produit
Description
Tarif

Pictogrammes

FlameExclamation mark

Mention d'avertissement

Danger

Mentions de danger

Classification des risques

Aquatic Chronic 3 - Desen. Expl. 2 - Eye Irrit. 2

Risques supp

Code de la classe de stockage

4.1A - Other explosive hazardous materials

Classe de danger pour l'eau (WGK)

WGK 1

Point d'éclair (°F)

Not applicable

Point d'éclair (°C)

Not applicable


Faites votre choix parmi les versions les plus récentes :

Certificats d'analyse (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

Si vous avez besoin d'assistance, veuillez contacter Service Clients

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Injectable gel scaffold based on biopolymer microspheres via an enzymatic reaction.
Huaping Tan et al.
Advanced healthcare materials, 3(11), 1769-1775 (2014-05-17)
M Zouhair Atassi et al.
Journal of neuroimmunology, 272(1-2), 29-34 (2014-05-21)
In cervical dystonia, injection of botulinum neurotoxin (BoNT) A or B into affected neck muscle reduces symptoms but may elicit anti-toxin antibodies (Abs) that block responsiveness to treatment. Previously, we localized the BoNT/A and BoNT/B sites that bind mouse or
Binu Jacob et al.
Journal of peptide science : an official publication of the European Peptide Society, 19(11), 700-707 (2013-10-10)
KR-12 (residues 18-29 of LL-37) was known to be the smallest peptide of human cathelicidin LL-37 possessing antimicrobial activity. In order to optimize α-helical short antimicrobial peptides having both antimicrobial and antiendotoxic activities without mammalian cell toxicity, we designed and
Mija Ahn et al.
Journal of medicinal chemistry, 58(1), 294-304 (2014-10-28)
Recent progress in the development of peptide-derived Polo-like kinase (Plk1) polo-box domain (PBD) inhibitors has led to the synthesis of multiple peptide ligands with high binding affinity and selectivity. However, few systematic analyses have been conducted to identify key Plk1
So Jeong Yoon et al.
Journal of bioscience and bioengineering, 118(4), 461-468 (2014-05-27)
Hydrogel has been used for regenerating myocardial infraction (MI) as a delivery vehicle for cells and growth factors. This study showed that injectable hyaluronic acid (HA)-based hydrogels alone would effectively regenerate the damaged infarcted heart tissue. We found that there

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique