Principaux documents
Z743203
Julabo® bath carrier trays for SW22/SW23
universal basic tray for assembling spring clamps
1 EA
352,00 €
352,00 €
Check Cart for Availability
Changer de vue
1 EA
352,00 €
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
352,00 €
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8970630
Description générale
Basic stainless steel tray for assembling Erlenmeyer flasks from 25 to 1000 mL, including spring clamps. Arrange springs to hold flasks in preferred arrangement. For use in shaking water bath models SW22, SW23.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.