Principaux documents
Z743136
Julabo® cryo-compact circulator
CF40, capacity 5.5 L, temperature -40-150 °C, precision: ±0.03, AC/DC input 230 V AC, Schuko plug
Synonyme(s) :
Julabo Cryostat
1 EA
6 380,00 €
6 380,00 €
Check Cart for Availability
Changer de vue
1 EA
6 380,00 €
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
6 380,00 €
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 9400340
Plage de mesure de la température
-40-150 °C, precision: ±0.03
Capacité
5.5 L
Description générale
The new CF series offers compact refrigerated/heating circulators. With small overall dimensions the units can even be placed on a very small surface or within a complex installation. All models offer a heating capacity of 2 kW and safety class 3 according to DIN12876-1. The maximum ambient temperature of +40°C as well as a ventilation air cooling allow installing the units very closely side by side with other units or directly under a fume hood. Furthermore all the models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. Wetted parts are made of high quality stainless steel or plastic. The instruments provide a reliable Microprocessor electronics with high temperature stability as well as warning and safety functions.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.