Z224197
Chem-FLEX™ tubing clamp
for tubing o.d., 1/4 ‑ 0.525 in.
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.35
Produits recommandés
Description
for tubing CHEM-FLEX 250
Caractéristiques
for tubing o.d. 1/4 ‑ 0.525 in.
Description générale
Noncorroding, lightweight nylon clamps secure CHEM-FLEX tubing to transfer needles and fittings.
Informations légales
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC