Principaux documents
Z169463
Supelco
Spectroline® E-Series lamp bulb
shortwave, output6 W
1 EA
48,30 €
48,30 €
Tarif69,00 €Économiser 30 %
Check Cart for Availability
Devis pour commande en gros
Changer de vue
1 EA
48,30 €
About This Item
Code UNSPSC :
41115711
Nomenclature NACRES :
NC.07
48,30 €
Tarif69,00 €Économiser 30 %
Check Cart for Availability
Devis pour commande en gros
Technique(s)
thin layer chromatography (TLC): suitable
Sortie
6 W
Application
Spectroline E-Series lamp bulb was used for illumination of trans-bis(trimethylammoniumhexyloxy)azobenzene dibromide solution to produce solution of its cis isomer†.[1]
Informations légales
Spectroline is a registered trademark of Spectronics, Corp.
Produit(s) apparenté(s)
Réf. du produit
Description
Tarif
Photocrosslinkable modified vegetable oil based resin for wood surface coating application
Mahendran, Arunjunai Raj, et al.
Progress in Organic Coatings, 74, 697-704 (2012)
F Pierce Hubbard et al.
Langmuir : the ACS journal of surfaces and colloids, 23(9), 4819-4829 (2007-03-27)
We report light and small-angle neutron scattering measurements that characterize microstructures formed in aqueous surfactant solutions (up to 1.0 wt % surfactant) containing mixtures of sodium dodecyl sulfate (SDS) and the light-sensitive bolaform surfactant, bis(trimethylammoniumhexyloxy)azobenzene dibromide (BTHA) as a function